Assay Details
| Gene Symbol: LOC101699691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021246864.1 | XP_021102523.1 | 3 - 4 | 405 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101699691| Gene Symbol: LOC101699691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021246864.1 | XP_021102523.1 | 3 - 4 | 405 | 73 | |