Assay Details
| Gene Symbol: Acvr1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835920.2 | XP_004835977.1 | 8 - 9 | 1337 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acvr1c| Gene Symbol: Acvr1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835920.2 | XP_004835977.1 | 8 - 9 | 1337 | 65 | |
Set Membership: |
|