Assay Details
| Gene Symbol: Maml2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838078.2 | XP_004838135.1 | 4 - 5 | 3705 | 70 | |
| XM_004838079.2 | XP_004838136.1 | 4 - 5 | 3573 | 70 | ||
More Information
Set Membership: |
|

Maml2
Genomic Map