Assay Details
Gene Symbol: Nrsn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004840618.2 | XP_004840675.1 | 1 - 2 | 79 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004840618.2 transcripts.
Set Membership: |
![]() ![]() |
