Assay Details
| Gene Symbol: Lipi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842198.2 | XP_004842255.1 | 1 - 2 | 131 | 66 | |
| XM_013071420.2 | XP_012926874.1 | 1 - 2 | 131 | 66 | ||
More Information
Set Membership: |
|

Lipi
Genomic Map