Assay Details
| Gene Symbol: Il10rb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842306.2 | XP_004842363.1 | 6 - 7 | 890 | 71 | |
| XM_013071306.2 | XP_012926760.1 | 6 - 7 | 987 | 71 | ||
More Information
Set Membership: |
|

Il10rb
Genomic Map