Assay Details
| Gene Symbol: LOC101721350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848050.2 | XP_004848107.1 | 15 - 16 | 2548 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101721350| Gene Symbol: LOC101721350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848050.2 | XP_004848107.1 | 15 - 16 | 2548 | 89 | |
Set Membership: |
|