Assay Details
| Gene Symbol: LOC101721350 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848050.2 | XP_004848107.1 | 5 - 6 | 1252 | 73 | |
| XM_021261051.1 | XP_021116710.1 | 5 - 6 | 1252 | 73 | ||
| XM_021261052.1 | XP_021116711.1 | 5 - 6 | 1252 | 73 | ||
More Information
Set Membership: |
|

LOC101721350
Genomic Map