Assay Details
Gene Symbol: Cd81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852102.3 | XP_004852159.1 | 5 - 6 | 688 | 66 | |
XM_013074899.2 | XP_012930353.1 | 5 - 6 | 442 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
