Assay Details
| Gene Symbol: Ccnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004852170.3 | XP_004852227.1 | 2 - 3 | 578 | 85 | |
| XM_021264461.1 | XP_021120120.1 | 2 - 3 | 583 | 85 | ||
More Information
Set Membership: |
|

Ccnd1
Genomic Map