Assay Details
| Gene Symbol: Parp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855624.2 | XP_004855681.1 | 6 - 7 | 1047 | 71 | |
| XM_004855625.2 | XP_004855682.1 | 5 - 6 | 857 | 71 | ||
| XM_004855626.2 | XP_004855683.1 | 5 - 6 | 871 | 71 | ||
| XM_013076254.2 | XP_012931708.1 | 6 - 7 | 1068 | 71 | ||
| XM_021237065.1 | XP_021092724.1 | 4 - 5 | 956 | 71 | ||
More Information
Set Membership: |
|

Parp16
Genomic Map