Assay Details
| Gene Symbol: Spag7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857320.3 | XP_004857377.1 | 4 - 5 | 463 | 66 | |
| XM_021238089.1 | XP_021093748.1 | 4 - 5 | 694 | 66 | ||
More Information
Set Membership: |
|

Spag7
Genomic Map