Assay Details
| Gene Symbol: LOC101710773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864918.3 | XP_004864975.1 | 4 - 5 | 574 | 66 | |
| XM_021244514.1 | XP_021100173.1 | 4 - 5 | 576 | 66 | ||
More Information
Set Membership: |
|

LOC101710773
Genomic Map