Assay Details
| Gene Symbol: S100z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870171.2 | XP_004870228.1 | 1 - 2 | 388 | 80 | |
| XM_021248572.1 | XP_021104231.1 | 2 - 3 | 665 | 80 | ||
More Information
Set Membership: |
|

S100z
Genomic Map