Assay Details
| Gene Symbol: Hlcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021256276.1 | XP_021111935.1 | 6 - 7 | 1936 | 58 | |
| XM_021256277.1 | XP_021111936.1 | 6 - 7 | 1795 | 58 | ||
More Information
Set Membership: |
|

Hlcs
Genomic Map