Assay Details
| Gene Symbol: LOC101716093 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859318.3 | XP_004859375.1 | 1 - 2 | 206 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004859318.3 transcripts.
Set Membership: |
|

LOC101716093
Genomic Map