Assay Details
| Gene Symbol: LOC101722810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252167.1 | XP_021107826.1 | 1 | 1619 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101722810| Gene Symbol: LOC101722810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252167.1 | XP_021107826.1 | 1 | 1619 | 100 | |
Set Membership: |
|