Assay Details
| Gene Symbol: LOC101703332 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862837.2 | XP_004862894.1 | 1 | 122 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004862837.2 transcripts.
Set Membership: |
|

LOC101703332
Genomic Map