Assay Details
| Gene Symbol: Bricd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021251326.1 | XP_021106985.1 | 1 - 2 | 617 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021251326.1 transcripts.
Set Membership: |
|

Bricd5
Genomic Map