Assay Details
| Gene Symbol: Tbca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870187.2 | XP_004870244.1 | 1 - 2 | 171 | 83 | |
| XM_021248566.1 | XP_021104225.1 | 1 - 2 | 172 | 83 | ||
More Information
Set Membership: |
|

Tbca
Genomic Map