Assay Details
| Gene Symbol: LOC101706939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021255668.1 | XP_021111327.1 | 2 | 825 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101706939| Gene Symbol: LOC101706939 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021255668.1 | XP_021111327.1 | 2 | 825 | 84 | |
Set Membership: |
|