Assay Details
| Gene Symbol: LOC110347628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252642.1 | XP_021108301.1 | 1 | 1316 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC110347628| Gene Symbol: LOC110347628 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021252642.1 | XP_021108301.1 | 1 | 1316 | 99 | |
Set Membership: |
|