Assay Details
| Gene Symbol: Eif1ax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866174.3 | XP_004866231.1 | 7 | 2514 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif1ax| Gene Symbol: Eif1ax | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866174.3 | XP_004866231.1 | 7 | 2514 | 114 | |
Set Membership: |
|