Assay Details
| Gene Symbol: Trappc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870995.2 | XP_004871052.1 | 5 | 1014 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trappc2| Gene Symbol: Trappc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870995.2 | XP_004871052.1 | 5 | 1014 | 122 | |
Set Membership: |
|