| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006932.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006932.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR22006932.hg.1 |
TC22000361.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007544.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007544.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007544.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007544.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007544.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007544.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007544.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007544.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007544.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007544.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007544.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2200007544.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200153501.hg.1 |
TC2200007544.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2200153501.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200153501.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2200153501.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2200153501.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007544.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007544.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007544.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007544.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007544.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007544.hg.1 |
ENST00000216115.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2200007544.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007544.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007544.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007544.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007544.hg.1 |
NM_001197.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2200007544.hg.1 |
NM_001197.4 |
Refseq |