Assay Details
| Gene Symbol: CA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001218.4 | NP_001209.1 | 2 - 3 | 497 | 69 | |
| NM_206925.2 | NP_996808.1 | 2 - 3 | 497 | 69 | ||
| NR_135511.1 | 2 - 3 | 553 | 69 | |||
| GenBank mRNA | AF037335.1 | - | 2 - 3 | 222 | 69 | |
| AF051882.1 | - | 2 - 3 | 114 | 69 | ||
| AK315769.1 | - | 2 - 3 | 263 | 69 | ||
| BC000278.1 | - | 2 - 3 | 219 | 3357439 | 69 | |
| BC011691.2 | - | 2 - 3 | 222 | 3344734 | 69 | |
| BC023981.1 | - | 2 - 3 | 237 | 4042589 | 69 | |
| BQ440047.1 | - | 2 - 3 | 481 | 69 | ||
| BT006656.1 | - | 2 - 3 | 107 | 69 | ||
| DA509400.1 | - | 2 - 3 | 497 | 69 | ||
| DA546357.1 | - | 2 - 3 | 432 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001541.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000560293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000560293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014507.hg.1 | ENST00000178638.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014507.hg.1 | ENST00000344366.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014507.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014507.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014507.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014507.hg.1 | TC15001541.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500171624.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171624.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171624.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500171624.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500171624.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171624.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500171624.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500171624.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500171624.hg.1 | TC1500009709.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000178638.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000344366.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000422263.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000558287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000560293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000560293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | ENST00000560666.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001218.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_001293642.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009709.hg.1 | NM_206925.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 158502 | - | - |

CA12
Genomic Map