Assay Details
| Gene Symbol: LY75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002349.3 | NP_002340.2 | 16 - 17 | 2462 | 127 | |
| GenBank mRNA | AB208915.1 | - | 10 - 11 | 1242 | 127 | |
| AF011333.1 | - | 16 - 17 | 2441 | 127 | ||
| AF064827.1 | - | 16 - 17 | 2388 | 127 | ||
| DB146492.1 | - | 1 - 2 | 65 | 127 | ||
| Gene Symbol: LY75-CD302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198759.1 | NP_001185688.1 | 16 - 17 | 2462 | 127 | |
| NM_001198760.1 | NP_001185689.1 | 16 - 17 | 2462 | 127 | ||
| GenBank mRNA | AY184222.1 | - | 16 - 17 | 2388 | 127 | |
| AY314006.1 | - | 16 - 17 | 2388 | 127 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LY75
Genomic Map