Assay Details
| Gene Symbol: PSMB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002800.4 | NP_002791.1 | 1 - 2 | 131 | 80 | |
| GenBank mRNA | AK303118.1 | - | 1 - 2 | 134 | 80 | |
| BC065513.1 | - | 1 - 2 | 86 | 5749985 | 80 | |
| BI832878.1 | - | 1 - 2 | 113 | 80 | ||
| BX641100.1 | - | 1 - 2 | 144 | 80 | ||
| CR541656.1 | - | 1 - 2 | 62 | 80 | ||
| X62741.1 | - | 1 - 2 | 77 | 80 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06004082.hg.1, TC6_apd_hap1000122.hg.1, TC6_cox_hap2000259.hg.1, TC6_dbb_hap3000226.hg.1, TC6_mann_hap4000190.hg.1, TC6_mcf_hap5000202.hg.1, TC6_qbl_hap6000215.hg.1, TC6_ssto_hap7000193.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein polyubiquitination
liver development
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
regulation of cellular amino acid metabolic process
muscle atrophy
anaphase-promoting complex-dependent catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
response to drug
proteasome-mediated ubiquitin-dependent protein catabolic process
response to alkaloid
regulation of mRNA stability
spleen development
thymus development
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
Wnt signaling pathway, planar cell polarity pathway
cellular response to electrical stimulus
cellular response to interleukin-1
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
cellular response to virus
response to benzene
regulation of cysteine-type endopeptidase activity
threonine-type endopeptidase activity
protein binding
proteasome binding
MAPK cascade
protein polyubiquitination
liver development
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
regulation of cellular amino acid metabolic process
muscle atrophy
anaphase-promoting complex-dependent catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
response to drug
proteasome-mediated ubiquitin-dependent protein catabolic process
response to alkaloid
regulation of mRNA stability
spleen development
thymus development
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
Wnt signaling pathway, planar cell polarity pathway
cellular response to electrical stimulus
cellular response to interleukin-1
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
cellular response to virus
response to benzene
regulation of cysteine-type endopeptidase activity
threonine-type endopeptidase activity
protein binding
proteasome binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001580.hg.1 | TC6_mcf_hap5000202.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001580.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004707.hg.1 | TC06004082.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004707.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004707.hg.1 | ENST00000395333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004707.hg.1 | ENST00000453265.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004707.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000679.hg.1 | TC6_apd_hap1000122.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000679.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000676.hg.1 | TC6_apd_hap1000122.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000676.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000677.hg.1 | TC6_apd_hap1000122.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_apd_hap1000677.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001788.hg.1 | TC6_cox_hap2000259.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001788.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001803.hg.1 | TC6_dbb_hap3000226.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001803.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004703.hg.1 | TC06004082.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004703.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004703.hg.1 | ENST00000453265.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004703.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001799.hg.1 | TC6_dbb_hap3000226.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001799.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001755.hg.1 | TC6_qbl_hap6000215.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001755.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001401.hg.1 | TC6_mann_hap4000190.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001401.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001611.hg.1 | TC6_ssto_hap7000193.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001611.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001399.hg.1 | TC6_mann_hap4000190.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001399.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004705.hg.1 | TC06004082.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004705.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004705.hg.1 | ENST00000395333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004705.hg.1 | ENST00000453265.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06004705.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001397.hg.1 | TC6_mann_hap4000190.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mann_hap4001397.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001753.hg.1 | TC6_qbl_hap6000215.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001753.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001613.hg.1 | TC6_ssto_hap7000193.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001613.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001790.hg.1 | TC6_cox_hap2000259.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001790.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001792.hg.1 | TC6_cox_hap2000259.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_cox_hap2001792.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001801.hg.1 | TC6_dbb_hap3000226.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_dbb_hap3001801.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001577.hg.1 | TC6_mcf_hap5000202.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001577.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001578.hg.1 | TC6_mcf_hap5000202.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_mcf_hap5001578.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001751.hg.1 | TC6_qbl_hap6000215.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_qbl_hap6001751.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001610.hg.1 | TC6_ssto_hap7000193.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR6_ssto_hap7001610.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198413.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198413.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198413.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198413.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198413.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198413.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198413.hg.1 | TC0600014110.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014110.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198415.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198415.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198415.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198415.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198415.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198415.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198415.hg.1 | TC0600014110.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600198417.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198417.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198417.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198417.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198417.hg.1 | ENST00000374859.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198417.hg.1 | ENST00000464863.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198417.hg.1 | ENST00000467593.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600198417.hg.1 | NM_002800.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600198417.hg.1 | TC0600014110.hg.1 | Clariom D TC |

PSMB9
Genomic Map