Assay Details
| Gene Symbol: SLC22A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB075951.1 | - | 8 - 9 | 2847 | 73 | |
| AK290787.1 | - | 9 - 10 | 1578 | 73 | ||
| BC039899.1 | - | 9 - 10 | 1555 | 5186229 | 73 | |
| X98333.1 | - | 9 - 10 | 1644 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000489644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000489644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000489644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000489644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | ENST00000498556.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600013775.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194988.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194988.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194988.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194988.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194988.hg.1 | ENST00000366953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194988.hg.1 | ENST00000486916.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194988.hg.1 | ENST00000491092.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600194988.hg.1 | NM_003058.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600194988.hg.1 | TC0600013775.hg.1 | Clariom D TC |

SLC22A2
Genomic Map