Assay Details
| Gene Symbol: SCN4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000334.4 | NP_000325.4 | 5 - 6 | 779 | 107 | |
| GenBank mRNA | AY212253.1 | - | 5 - 6 | 702 | 107 | |
| M81758.1 | - | 5 - 6 | 779 | 107 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001793.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
potassium ion transport
sodium ion transport
muscle contraction
choline transport
neuronal action potential
regulation of ion transmembrane transport
sodium ion transmembrane transport
regulation of postsynaptic membrane potential
membrane depolarization during action potential
voltage-gated sodium channel activity
potassium ion transport
sodium ion transport
muscle contraction
choline transport
neuronal action potential
regulation of ion transmembrane transport
sodium ion transmembrane transport
regulation of postsynaptic membrane potential
membrane depolarization during action potential
voltage-gated sodium channel activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17023964.hg.1 | ENST00000435607.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023964.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023964.hg.1 | TC17001793.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000581514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000581514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000581514.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000584310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000584310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000584310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | ENST00000584310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700192267.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700192267.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700192267.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700192267.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700192267.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700192267.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700192267.hg.1 | TC1700011431.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000435607.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000578147.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000581514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000581514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000581514.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000584310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000584310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000584310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | ENST00000584310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011431.hg.1 | NM_000334.4 | Refseq |

SCN4A
Genomic Map