Assay Details
| Gene Symbol: AQP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000486.5 | NP_000477.1 | 1 - 2 | 447 | 98 | |
| GenBank mRNA | AK225940.1 | - | 1 - 2 | 447 | 98 | |
| BC042496.1 | - | 1 - 2 | 442 | 5186409 | 98 | |
| CR542024.1 | - | 1 - 2 | 357 | 98 | ||
| S73196.1 | - | 1 - 2 | 427 | 98 | ||
| S73197.1 | - | 1 - 2 | 427 | 98 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000383.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
renal water homeostasis
renal water transport
water transport
excretion
cellular water homeostasis
glycerol transport
ion transmembrane transport
cellular response to water deprivation
cellular response to copper ion
cellular response to mercury ion
metanephric collecting duct development
water transmembrane transporter activity
glycerol transmembrane transporter activity
water channel activity
glycerol channel activity
renal water homeostasis
renal water transport
water transport
excretion
cellular water homeostasis
glycerol transport
ion transmembrane transport
cellular response to water deprivation
cellular response to copper ion
cellular response to mercury ion
metanephric collecting duct development
water transmembrane transporter activity
glycerol transmembrane transporter activity
water channel activity
glycerol channel activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12004763.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004763.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004763.hg.1 | TC12000383.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004764.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004764.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004764.hg.1 | TC12000383.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004761.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004761.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004761.hg.1 | TC12000383.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004762.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004762.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12004762.hg.1 | TC12000383.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200153632.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153632.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153632.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153632.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153632.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153632.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153632.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153632.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153632.hg.1 | TC1200007590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200153634.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153634.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153634.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153634.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153634.hg.1 | TC1200007590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200153636.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153636.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153636.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153636.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153636.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153636.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153636.hg.1 | TC1200007590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200153628.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153628.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153628.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153628.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153628.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153628.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153628.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153628.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153628.hg.1 | TC1200007590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200153633.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153633.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153633.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153633.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153633.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153633.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153633.hg.1 | TC1200007590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200007590.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153629.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153629.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153629.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153629.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153629.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153629.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153629.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153629.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153629.hg.1 | TC1200007590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200153637.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153637.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153637.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153637.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153637.hg.1 | TC1200007590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200153630.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153630.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153630.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153630.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153630.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153630.hg.1 | ENST00000550862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153630.hg.1 | ENST00000551526.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153630.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153630.hg.1 | TC1200007590.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200153635.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153635.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153635.hg.1 | ENST00000199280.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200153635.hg.1 | NM_000486.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200153635.hg.1 | TC1200007590.hg.1 | Clariom D TC |

AQP2
Genomic Map