Assay Details
| Gene Symbol: IL12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000882.3 | NP_000873.2 | 2 - 3 | 486 | 67 | |
| GenBank mRNA | AF101062.1 | - | 2 - 3 | 265 | 67 | |
| AF180562.1 | - | 2 - 3 | 324 | 67 | ||
| BC104982.1 | - | 2 - 3 | 418 | 8143985 | 67 | |
| BC104984.1 | - | 2 - 3 | 418 | 8143987 | 67 | |
| BG702253.1 | - | 2 - 3 | 579 | 67 | ||
| M65271.1 | - | 2 - 3 | 332 | 67 | ||
| M65291.1 | - | 2 - 3 | 366 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000870.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of T cell mediated cytotoxicity
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
immune response
cell cycle arrest
response to virus
response to UV-B
cell migration
response to lipopolysaccharide
negative regulation of interleukin-17 production
positive regulation of interferon-gamma production
positive regulation of natural killer cell activation
positive regulation of mononuclear cell proliferation
positive regulation of smooth muscle cell apoptotic process
positive regulation of T cell proliferation
positive regulation of tyrosine phosphorylation of Stat4 protein
defense response to protozoan
positive regulation of T cell differentiation
positive regulation of cell adhesion
positive regulation of natural killer cell mediated cytotoxicity
negative regulation of smooth muscle cell proliferation
positive regulation of lymphocyte proliferation
defense response to Gram-positive bacterium
positive regulation of NK T cell activation
cellular response to lipopolysaccharide
extrinsic apoptotic signaling pathway
cellular response to virus
positive regulation of dendritic cell chemotaxis
cytokine activity
interleukin-12 receptor binding
protein binding
growth factor activity
interleukin-12 beta subunit binding
interleukin-27 binding
protein heterodimerization activity
positive regulation of T cell mediated cytotoxicity
positive regulation of natural killer cell mediated cytotoxicity directed against tumor cell target
immune response
cell cycle arrest
response to virus
response to UV-B
cell migration
response to lipopolysaccharide
negative regulation of interleukin-17 production
positive regulation of interferon-gamma production
positive regulation of natural killer cell activation
positive regulation of mononuclear cell proliferation
positive regulation of smooth muscle cell apoptotic process
positive regulation of T cell proliferation
positive regulation of tyrosine phosphorylation of Stat4 protein
defense response to protozoan
positive regulation of T cell differentiation
positive regulation of cell adhesion
positive regulation of natural killer cell mediated cytotoxicity
negative regulation of smooth muscle cell proliferation
positive regulation of lymphocyte proliferation
defense response to Gram-positive bacterium
positive regulation of NK T cell activation
cellular response to lipopolysaccharide
extrinsic apoptotic signaling pathway
cellular response to virus
positive regulation of dendritic cell chemotaxis
cytokine activity
interleukin-12 receptor binding
protein binding
growth factor activity
interleukin-12 beta subunit binding
interleukin-27 binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480088.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03015493.hg.1 | TC03000870.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03015493.hg.1 | ENST00000305579.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03015493.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03015493.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300167043.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300167043.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300167043.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300167043.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300167043.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300167043.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300167043.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300167043.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300167043.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300167043.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300167043.hg.1 | TC0300009328.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000305579.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000466512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000468862.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480088.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000480787.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | ENST00000496308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009328.hg.1 | NM_000882.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 103164 | - | - |

IL12A
Genomic Map