Assay Details
| Gene Symbol: NCKAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013436.4 | NP_038464.1 | 1 - 2 | 867 | 70 | |
| XM_006712201.3 | XP_006712264.1 | 1 - 2 | 361 | 70 | ||
| GenBank mRNA | AB014509.1 | - | 1 - 2 | 127 | 70 | |
| AK292914.1 | - | 1 - 2 | 357 | 70 | ||
| BC015025.1 | - | 1 - 2 | 379 | 3907646 | 70 | |
| BT007033.1 | - | 1 - 2 | 108 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

NCKAP1
Genomic Map