| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000308744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000308744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000308744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000308744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000308744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000308744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000308744.10 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000344413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000344413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000344413.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000358749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000358749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000358749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000358749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000358749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000358749.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000395999.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000395999.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000395999.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000395999.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000395999.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000395999.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000395999.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000395999.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000506359.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000506359.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000506359.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
ENST00000506359.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_001144756.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_001144756.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_001144756.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_001144756.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_001144756.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_001144756.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_001144756.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_001144756.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_001144756.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_001144756.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_004885.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_004885.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_004885.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_004885.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_004885.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_004885.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_004885.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_004885.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_053036.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_053036.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_053036.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_053036.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_053036.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_053036.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_053036.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_053036.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007806.hg.1 |
NM_053036.2 |
Refseq |