Assay Details
| Gene Symbol: LIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002309.4 | NP_002300.1 | 1 - 2 | 179 | 66 | |
| GenBank mRNA | AK315310.1 | - | 1 - 2 | 127 | 66 | |
| BC069540.1 | - | 1 - 2 | 73 | 7262225 | 66 | |
| BC093733.1 | - | 1 - 2 | 71 | 7939578 | 66 | |
| BC093735.1 | - | 1 - 2 | 71 | 7939580 | 66 | |
| CN409458.1 | - | 1 - 2 | 179 | 66 | ||
| X13967.1 | - | 1 - 2 | 67 | 66 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK315310.1,BC069540.1,BC093733.1,BC093735.1,NM_002309.4,X13967.1 transcripts.
Set Membership: |
|

LIF
Genomic Map