Assay Details
| Gene Symbol: SPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003123.4 | NP_003114.1 | 1 - 2 | 102 | 70 | |
| GenBank mRNA | AK292626.1 | - | 1 - 2 | 74 | 70 | |
| AK313750.1 | - | 1 - 2 | 74 | 70 | ||
| DB108930.1 | - | 1 - 2 | 102 | 70 | ||
| J04536.1 | - | 1 - 2 | 62 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16002068.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK292626.1,AK313750.1,J04536.1,NM_003123.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
response to protozoan
negative regulation of type IV hypersensitivity
positive regulation of protein phosphorylation
chemotaxis
immune response
cellular defense response
negative regulation of cell adhesion
establishment or maintenance of cell polarity
signal transduction
cell surface receptor signaling pathway
T cell costimulation
positive regulation of T cell proliferation
negative regulation of T cell proliferation
positive regulation of tumor necrosis factor biosynthetic process
defense response to bacterium
negative thymic T cell selection
regulation of defense response to virus
leukocyte migration
apoptotic signaling pathway
transmembrane signaling receptor activity
response to protozoan
negative regulation of type IV hypersensitivity
positive regulation of protein phosphorylation
chemotaxis
immune response
cellular defense response
negative regulation of cell adhesion
establishment or maintenance of cell polarity
signal transduction
cell surface receptor signaling pathway
T cell costimulation
positive regulation of T cell proliferation
negative regulation of T cell proliferation
positive regulation of tumor necrosis factor biosynthetic process
defense response to bacterium
negative thymic T cell selection
regulation of defense response to virus
leukocyte migration
apoptotic signaling pathway
transmembrane signaling receptor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000561857.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004596.hg.1 | TC16002068.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004596.hg.1 | ENST00000360121.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004596.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004598.hg.1 | TC16002068.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004598.hg.1 | ENST00000360121.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004598.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004598.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16004598.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600190057.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190057.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190057.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190057.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190057.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600190057.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600190057.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190057.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190057.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190057.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190057.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600190057.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600190057.hg.1 | TC1600011374.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600190055.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190055.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190055.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190055.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600190055.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190055.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190055.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190055.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600190055.hg.1 | TC1600011374.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600190054.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190054.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190054.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600190054.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190054.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190054.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600190054.hg.1 | TC1600011374.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600190056.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190056.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190056.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190056.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190056.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600190056.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600190056.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190056.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190056.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190056.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600190056.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600190056.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600190056.hg.1 | TC1600011374.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000360121.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000395389.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000436527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000561857.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | ENST00000563039.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_001030288.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_003123.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011374.hg.1 | NM_003123.5 | Refseq |

SPN
Genomic Map