Assay Details
| Gene Symbol: ITGA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001303040.1 | NP_001289969.1 | 1 - 2 | 231 | 64 | |
| NM_003637.4 | NP_003628.2 | 1 - 2 | 231 | 64 | ||
| XM_005277436.3 | XP_005277493.1 | 1 - 2 | 92 | 64 | ||
| GenBank mRNA | AF074015.1 | - | 1 - 2 | 77 | 64 | |
| AF112345.1 | - | 1 - 2 | 132 | 64 | ||
| AK304155.1 | - | 1 - 2 | 87 | 64 | ||
| AK304183.1 | - | 1 - 2 | 87 | 64 | ||
| AK314255.1 | - | 1 - 2 | 80 | 64 | ||
| AY358325.1 | - | 1 - 2 | 189 | 64 | ||
| BM311010.1 | - | 1 - 2 | 234 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001095.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK304155.1,AK304183.1,NM_001303040.1,XM_005277436.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000369304.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000468261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000481236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000481236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000481236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000481236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | ENST00000539363.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303040.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_001303041.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100015594.hg.1 | NM_003637.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 116215 | - | - |

ITGA10
Genomic Map