| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02009798.hg.1 |
TC02000619.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02009798.hg.1 |
ENST00000332549.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02009798.hg.1 |
ENST00000393414.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02009798.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02009798.hg.1 |
ENST00000485335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02009798.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000457817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000457817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000457817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000457817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000457817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000457817.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000464994.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000464994.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000464994.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000464994.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000464994.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000474085.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000474085.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000474085.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000474085.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000474085.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000482658.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000482658.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000482658.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000485335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000485335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000485335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000485335.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000493749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000493749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000493749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000493749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000493749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
ENST00000493749.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
ENST00000474085.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
ENST00000485335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
ENST00000474085.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
ENST00000485335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160463.hg.1 |
TC0200008663.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000332549.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000393414.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000457817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000457817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000457817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000457817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000457817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000457817.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000464994.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000464994.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000464994.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000464994.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000464994.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000474085.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000474085.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000474085.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000474085.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000474085.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000482658.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000482658.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000482658.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000485335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000485335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000485335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000485335.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000493749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000493749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000493749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000493749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000493749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
ENST00000493749.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NM_004633.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008663.hg.1 |
NR_048564.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160465.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160465.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160465.hg.1 |
ENST00000441002.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200160465.hg.1 |
NM_001261419.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200160465.hg.1 |
TC0200008663.hg.1 |
Clariom D TC |