Assay Details
| Gene Symbol: PTPRCAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005608.2 | NP_005599.1 | 1 - 2 | 71 | 92 | |
| GenBank mRNA | AK312022.1 | - | 1 - 2 | 71 | 92 | |
| BC113859.1 | - | 1 - 2 | 70 | 40030438 | 92 | |
| BC114467.1 | - | 1 - 2 | 70 | 40030437 | 92 | |
| X81422.1 | - | 1 - 2 | 71 | 92 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11003489.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK312022.1,BC113859.1,BC114467.1,NM_005608.2,X81422.1 transcripts.
Set Membership: |
|

PTPRCAP
Genomic Map