Assay Details
| Gene Symbol: RIPK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006871.3 | NP_006862.2 | 5 - 6 | 877 | 70 | |
| GenBank mRNA | AF156884.1 | - | 5 - 6 | 667 | 70 | |
| AK075275.1 | - | 5 - 6 | 831 | 70 | ||
| AY453693.1 | - | 5 - 6 | 667 | 70 | ||
| BC062584.1 | - | 5 - 6 | 808 | 5213027 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14002317.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of T cell mediated cytotoxicity
regulation of adaptive immune response
cellular protein modification process
signal transduction
I-kappaB kinase/NF-kappaB signaling
positive regulation of phosphatase activity
activation of protein kinase activity
regulation of interferon-gamma production
T cell differentiation in thymus
NIK/NF-kappaB signaling
T cell homeostasis
regulation of activated T cell proliferation
protein autophosphorylation
lymph node development
spleen development
thymus development
positive regulation of NF-kappaB transcription factor activity
protein homooligomerization
protein heterooligomerization
positive regulation of ligase activity
positive regulation of oxidoreductase activity
positive regulation of necroptotic process
regulation of activation-induced cell death of T cells
necroptotic process
positive regulation of protein deacetylation
apoptotic signaling pathway
amyloid fibril formation
positive regulation of reactive oxygen species metabolic process
regulation of CD8-positive, alpha-beta cytotoxic T cell extravasation
positive regulation of intrinsic apoptotic signaling pathway
transcription coactivator activity
protein kinase activity
protein serine/threonine kinase activity
NF-kappaB-inducing kinase activity
protein binding
ATP binding
protein complex binding
identical protein binding
regulation of T cell mediated cytotoxicity
regulation of adaptive immune response
cellular protein modification process
signal transduction
I-kappaB kinase/NF-kappaB signaling
positive regulation of phosphatase activity
activation of protein kinase activity
regulation of interferon-gamma production
T cell differentiation in thymus
NIK/NF-kappaB signaling
T cell homeostasis
regulation of activated T cell proliferation
protein autophosphorylation
lymph node development
spleen development
thymus development
positive regulation of NF-kappaB transcription factor activity
protein homooligomerization
protein heterooligomerization
positive regulation of ligase activity
positive regulation of oxidoreductase activity
positive regulation of necroptotic process
regulation of activation-induced cell death of T cells
necroptotic process
positive regulation of protein deacetylation
apoptotic signaling pathway
amyloid fibril formation
positive regulation of reactive oxygen species metabolic process
regulation of CD8-positive, alpha-beta cytotoxic T cell extravasation
positive regulation of intrinsic apoptotic signaling pathway
transcription coactivator activity
protein kinase activity
protein serine/threonine kinase activity
NF-kappaB-inducing kinase activity
protein binding
ATP binding
protein complex binding
identical protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14011184.hg.1 | TC14002317.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14011184.hg.1 | ENST00000216274.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14011184.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554569.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400181982.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400181982.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400181982.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400181982.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400181982.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400181982.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400181982.hg.1 | TC1400010727.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000216274.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554338.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554569.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000554756.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557253.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557624.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | ENST00000557662.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010727.hg.1 | NM_006871.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 140702 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 235224 | - | - |

RIPK3
Genomic Map