Assay Details
| Gene Symbol: SSPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005086.4 | NP_005077.2 | 1 - 2 | 382 | 73 | |
| GenBank mRNA | AF016028.2 | - | 1 - 2 | 282 | 73 | |
| AK314353.1 | - | 1 - 2 | 371 | 73 | ||
| AL136756.1 | - | 1 - 2 | 366 | 73 | ||
| BC062299.1 | - | 1 - 2 | 362 | 6376868 | 73 | |
| EU433933.1 | - | 1 - 2 | 282 | 73 | ||
| X89105.1 | - | 1 - 2 | 204 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12003291.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12003120.hg.1 | TC12003291.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003120.hg.1 | ENST00000242729.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003120.hg.1 | ENST00000441067.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003120.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003120.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003122.hg.1 | TC12003291.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003122.hg.1 | ENST00000242729.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003122.hg.1 | ENST00000422622.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003122.hg.1 | ENST00000441067.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003122.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003122.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003122.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003118.hg.1 | TC12003291.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003118.hg.1 | ENST00000242729.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003118.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003119.hg.1 | TC12003291.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003119.hg.1 | ENST00000242729.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003119.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12003119.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199574.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199574.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199574.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199574.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199574.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199574.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199574.hg.1 | TC1200012610.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200199576.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199576.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199576.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199576.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199576.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199576.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199576.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199576.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199576.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199576.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199576.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199576.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199576.hg.1 | TC1200012610.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200199573.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199573.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199573.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199573.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199573.hg.1 | TC1200012610.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200199575.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199575.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199575.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199575.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199575.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199575.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199575.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199575.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199575.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199575.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199575.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199575.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199575.hg.1 | TC1200012610.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000242729.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000422622.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000535504.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | ENST00000544231.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_001135823.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012610.hg.1 | NM_005086.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 155357 | - | - |

SSPN
Genomic Map