Assay Details
| Gene Symbol: TLE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144761.1 | NP_001138233.1 | 5 - 6 | 356 | 58 | |
| NM_001144762.1 | NP_001138234.1 | 4 - 5 | 274 | 58 | ||
| NM_001300846.1 | NP_001287775.1 | 4 - 5 | 530 | 58 | ||
| NM_003260.4 | NP_003251.2 | 4 - 5 | 496 | 58 | ||
| XM_006722864.2 | XP_006722927.1 | 4 - 5 | 438 | 58 | ||
| XM_011528230.1 | XP_011526532.1 | 4 - 5 | 438 | 58 | ||
| GenBank mRNA | AK293407.1 | - | 4 - 5 | 276 | 58 | |
| AK303102.1 | - | 4 - 5 | 372 | 58 | ||
| AK308137.1 | - | 5 - 6 | 356 | 58 | ||
| AK308608.1 | - | 4 - 5 | 248 | 58 | ||
| AK312285.1 | - | 4 - 5 | 342 | 58 | ||
| BC017364.2 | - | 4 - 5 | 353 | 3925626 | 58 | |
| M99436.1 | - | 4 - 5 | 258 | 58 | ||
More Information
Set Membership: |
|

TLE2
Genomic Map