Assay Details
| Gene Symbol: PIR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018109.2 | NP_001018119.1 | 4 - 5 | 722 | 79 | |
| NM_003662.3 | NP_003653.1 | 4 - 5 | 756 | 79 | ||
| GenBank mRNA | AK309656.1 | - | 4 - 5 | 438 | 79 | |
| BC002517.2 | - | 4 - 5 | 370 | 3140037 | 79 | |
| BT019583.1 | - | 3 - 4 | 271 | 79 | ||
| BT019584.1 | - | 3 - 4 | 271 | 79 | ||
| CR541822.1 | - | 3 - 4 | 271 | 79 | ||
| DT217415.1 | - | 4 - 5 | 500 | 79 | ||
| Y07867.1 | - | 4 - 5 | 475 | 79 | ||
| Y07868.1 | - | 4 - 5 | 501 | 79 | ||
| Gene Symbol: PIR-FIGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037859.2 | 3 - 4 | 323 | 79 | ||
| GenBank mRNA | AX740234.1 | - | 3 - 4 | 1031 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X002327.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011729.hg.1 | TC0X002327.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011729.hg.1 | ENST00000380420.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011729.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011729.hg.1 | ENST00000476381.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011729.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X011729.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000476381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000476381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000476381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000476381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000476381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000484433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000484433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000484433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000484433.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000492432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000492432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000492432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000492432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000492432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | ENST00000492432.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00178596.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00178596.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00178596.hg.1 | ENST00000476381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00178596.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00178596.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00178596.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00178596.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00178596.hg.1 | ENST00000476381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00178596.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00178596.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00178596.hg.1 | TC0X00011355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00178620.hg.1 | NR_037859.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00178620.hg.1 | NR_037859.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00178620.hg.1 | TC0X00011356.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380420.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000380421.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000471725.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000476381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000476381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000476381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000476381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000476381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000484433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000484433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000484433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000484433.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000492432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000492432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000492432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000492432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000492432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | ENST00000492432.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_001018109.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011355.hg.1 | NM_003662.3 | Refseq |

PIR
Genomic Map