Assay Details
| Gene Symbol: TP63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114978.1 | NP_001108450.1 | 3 - 4 | 417 | 75 | |
| NM_001114979.1 | NP_001108451.1 | 3 - 4 | 417 | 75 | ||
| NM_003722.4 | NP_003713.3 | 3 - 4 | 417 | 75 | ||
| XM_005247843.3 | XP_005247900.1 | 3 - 4 | 864 | 75 | ||
| XM_005247844.4 | XP_005247901.1 | 3 - 4 | 580 | 75 | ||
| XM_011513251.2 | XP_011511553.1 | 4 - 5 | 458 | 75 | ||
| XM_011513252.2 | XP_011511554.1 | 3 - 4 | 390 | 75 | ||
| GenBank mRNA | AB016072.1 | - | 3 - 4 | 355 | 75 | |
| AB016073.1 | - | 3 - 4 | 355 | 75 | ||
| AF075428.1 | - | 2 - 3 | 211 | 75 | ||
| AF075430.1 | - | 2 - 3 | 211 | 75 | ||
| AF075432.1 | - | 2 - 3 | 211 | 75 | ||
| AF116771.1 | - | 3 - 4 | 355 | 75 | ||
| BC039815.1 | - | 3 - 4 | 417 | 5552611 | 75 | |
| KU178435.1 | - | 3 - 4 | 328 | 75 | ||
| KU178436.1 | - | 3 - 4 | 328 | 75 | ||
| Y16961.1 | - | 3 - 4 | 355 | 75 | ||
More Information
Set Membership: |
|

TP63
Genomic Map