Assay Details
| Gene Symbol: MBD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003927.4 | NP_003918.1 | 3 - 4 | 1074 | 67 | |
| GenBank mRNA | AF039701.1 | - | 2 - 3 | 212 | 67 | |
| AF072242.1 | - | 3 - 4 | 1074 | 67 | ||
| BC032638.1 | - | 3 - 4 | 915 | 5496721 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC18000521.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
chromatin silencing at rDNA
methylation-dependent chromatin silencing
transcription, DNA-templated
positive regulation of Wnt signaling pathway
regulation of cell proliferation
maternal behavior
ATP-dependent chromatin remodeling
cellular protein complex assembly
negative regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
satellite DNA binding
mRNA binding
protein binding
methyl-CpG binding
protein domain specific binding
nucleosomal DNA binding
siRNA binding
C2H2 zinc finger domain binding
negative regulation of transcription from RNA polymerase II promoter
chromatin silencing at rDNA
methylation-dependent chromatin silencing
transcription, DNA-templated
positive regulation of Wnt signaling pathway
regulation of cell proliferation
maternal behavior
ATP-dependent chromatin remodeling
cellular protein complex assembly
negative regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
satellite DNA binding
mRNA binding
protein binding
methyl-CpG binding
protein domain specific binding
nucleosomal DNA binding
siRNA binding
C2H2 zinc finger domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR18005422.hg.1 | ENST00000256429.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005422.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR18005422.hg.1 | TC18000521.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000398398.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000398398.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000398398.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000398398.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000398398.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000398398.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000578272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000578272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000578272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000578272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000578272.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000579025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000579025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000579025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000583046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000583046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000583046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000583046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | ENST00000583046.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_015832.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_015832.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_015832.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_015832.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_015832.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_015832.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1800008730.hg.1 | NM_015832.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800159864.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159864.hg.1 | ENST00000578272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159864.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800159864.hg.1 | TC1800008730.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800159864.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159864.hg.1 | ENST00000578272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1800159864.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000256429.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000398398.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000398398.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000398398.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000398398.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000398398.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000398398.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000578272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000578272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000578272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000578272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000578272.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000579025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000579025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000579025.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000583046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000583046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000583046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000583046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | ENST00000583046.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_003927.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_015832.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_015832.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_015832.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_015832.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_015832.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_015832.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1800008730.hg.1 | NM_015832.4 | Refseq |

MBD2
Genomic Map