Assay Details
| Gene Symbol: C14orf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004894.2 | NP_004885.1 | 1 - 2 | 99 | 90 | |
| GenBank mRNA | AF054175.1 | - | 1 - 2 | 62 | 90 | |
| AF116639.1 | - | 95 | 90 | |||
| AK309355.1 | - | 1 - 2 | 60 | 90 | ||
| AK309499.1 | - | 1 - 2 | 62 | 90 | ||
| AK312099.1 | - | 1 - 2 | 60 | 90 | ||
| BC000429.2 | - | 1 - 2 | 51 | 2819801 | 90 | |
| BC001944.1 | - | 1 - 2 | 51 | 2819801 | 90 | |
| DA526965.1 | - | 1 - 2 | 99 | 90 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001547.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF054175.1,AF116639.1,AK312099.1,BC000429.2,BC001944.1,NM_004894.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14017370.hg.1 | ENST00000286953.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017370.hg.1 | ENST00000414262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017370.hg.1 | ENST00000553430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017370.hg.1 | ENST00000554713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017370.hg.1 | ENST00000554880.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017370.hg.1 | ENST00000555030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017370.hg.1 | ENST00000557040.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017370.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017370.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017370.hg.1 | TC14001547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017381.hg.1 | ENST00000286953.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017381.hg.1 | ENST00000553430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017381.hg.1 | ENST00000553449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017381.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017381.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017381.hg.1 | TC14001547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017385.hg.1 | ENST00000286953.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017385.hg.1 | ENST00000553430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017385.hg.1 | ENST00000553449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017385.hg.1 | ENST00000554713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017385.hg.1 | ENST00000555030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017385.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017385.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017385.hg.1 | TC14001547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014215.hg.1 | ENST00000436385.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017384.hg.1 | ENST00000286953.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017384.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017384.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017384.hg.1 | TC14001547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017382.hg.1 | ENST00000286953.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017382.hg.1 | ENST00000553449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017382.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017382.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017382.hg.1 | TC14001547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017387.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017387.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017387.hg.1 | TC14001547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000553449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554528.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017386.hg.1 | ENST00000286953.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017386.hg.1 | ENST00000553430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017386.hg.1 | ENST00000553449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017386.hg.1 | ENST00000554713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017386.hg.1 | ENST00000555030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017386.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017386.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017386.hg.1 | TC14001547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0800008432.hg.1 | ENST00000528550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017383.hg.1 | ENST00000286953.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017383.hg.1 | ENST00000414262.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017383.hg.1 | ENST00000553430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017383.hg.1 | ENST00000553449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017383.hg.1 | ENST00000554713.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017383.hg.1 | ENST00000555030.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017383.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017383.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14017383.hg.1 | TC14001547.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100011968.hg.1 | ENST00000531740.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000554528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000554528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400176426.hg.1 | TC1400010339.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800008432.hg.1 | ENST00000528550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000286953.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000414262.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553430.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000553449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554528.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554713.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000554880.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000555030.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557040.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | ENST00000557260.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_001127393.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010339.hg.1 | NM_004894.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014215.hg.1 | ENST00000436385.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100011968.hg.1 | ENST00000531740.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 207683 | - | - |

C14orf2
Genomic Map