| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007777.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007777.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007777.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007777.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007777.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0400007777.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005205.hg.1 |
TC04000382.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005205.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005205.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005207.hg.1 |
TC04000382.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005207.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005207.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005204.hg.1 |
TC04000382.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005204.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005204.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005206.hg.1 |
TC04000382.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005206.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04005206.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400153069.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400153069.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400153069.hg.1 |
TC0400007777.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007777.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0400007777.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007777.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007777.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007777.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007777.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0400007777.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400153074.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400153074.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400153074.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400153074.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400153074.hg.1 |
TC0400007777.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400153073.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400153073.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400153073.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400153073.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400153073.hg.1 |
TC0400007777.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400153072.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400153072.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400153072.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400153072.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400153072.hg.1 |
TC0400007777.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400153070.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400153070.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400153070.hg.1 |
ENST00000304954.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400153070.hg.1 |
NM_005212.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400153070.hg.1 |
TC0400007777.hg.1 |
Clariom D TC |