Assay Details
| Gene Symbol: LIPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005357.3 | NP_005348.2 | 1 - 2 | 1162 | 67 | |
| XM_005258937.3 | XP_005258994.1 | 1 - 2 | 1157 | 67 | ||
| GenBank mRNA | AK292482.1 | - | 1 - 2 | 1142 | 67 | |
| BC029961.1 | - | 1 - 2 | 1149 | 67 | ||
| BC070041.1 | - | 1 - 2 | 1162 | 5296155 | 67 | |
| U40002.1 | - | 1 - 2 | 1162 | 67 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001573.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
cholesterol metabolic process
lipid catabolic process
triglyceride catabolic process
long-chain fatty acid catabolic process
diacylglycerol catabolic process
triglyceride lipase activity
protein binding
protein kinase binding
hormone-sensitive lipase activity
protein phosphorylation
cholesterol metabolic process
lipid catabolic process
triglyceride catabolic process
long-chain fatty acid catabolic process
diacylglycerol catabolic process
triglyceride lipase activity
protein binding
protein kinase binding
hormone-sensitive lipase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19021449.hg.1 | ENST00000244289.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021449.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021449.hg.1 | TC19001573.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000601189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000601189.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021451.hg.1 | ENST00000244289.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021451.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19021451.hg.1 | TC19001573.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000597620.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000599918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000600224.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000601189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000601189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | ENST00000602000.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010802.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900189409.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189409.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189409.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189409.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189409.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900189409.hg.1 | TC1900010802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900189409.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189409.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189409.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189409.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189409.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900189410.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189410.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189410.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189410.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189410.hg.1 | ENST00000601189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189410.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900189410.hg.1 | TC1900010802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900189410.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189410.hg.1 | ENST00000597001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189410.hg.1 | ENST00000599211.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189410.hg.1 | ENST00000599783.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189410.hg.1 | ENST00000601189.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189410.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900189417.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189417.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900189417.hg.1 | TC1900010802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900189417.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189417.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900189416.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189416.hg.1 | NM_005357.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900189416.hg.1 | TC1900010802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900189416.hg.1 | ENST00000244289.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900189416.hg.1 | NM_005357.3 | Refseq |
Citations (1)
Sex-specific alterations in mRNA level of key lipid metabolism enzymes in skeletal muscle of overweight and obese subjects following endurance exercise.
- Authors
- Smith IJ, Huffman KM, Durheim MT, Duscha BD, Kraus WE
- Journal
- Physiol Genomics - Page No 149 (Mon Feb 02 00:00:00 EST 2009)
- Volume (Year)
- 36 (2009)
- Product Usage
- -

LIPE
Genomic Map