Assay Details
Gene Symbol: ZNF354A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005649.2 | NP_005640.2 | 2 - 3 | 235 | 84 | |
XM_017009791.1 | XP_016865280.1 | 2 - 3 | 140 | 84 | ||
GenBank mRNA | AF116030.1 | - | 2 - 3 | 241 | 84 | |
AK290711.1 | - | 2 - 3 | 130 | 84 | ||
BC047105.1 | - | 2 - 3 | 192 | 5312219 | 84 | |
D89928.1 | - | 2 - 3 | 189 | 84 | ||
EU794664.1 | - | 2 - 3 | 235 | 84 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC05002126.hg.1
Additional Information:
This assay is in the 5'UTR region of the AF116030.1,AK290711.1,BC047105.1,D89928.1,EU794664.1,NM_005649.2,XM_017009791.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
response to hypoxia
kidney development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
nucleolar fragmentation
sensory perception of sound
response to folic acid
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
response to hypoxia
kidney development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
nucleolar fragmentation
sensory perception of sound
response to folic acid
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR05029602.hg.1 | TC05002126.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05029602.hg.1 | ENST00000335815.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05029602.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029605.hg.1 | TC05002126.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05029605.hg.1 | ENST00000335815.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05029605.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029607.hg.1 | TC05002126.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05029607.hg.1 | ENST00000335815.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05029607.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029601.hg.1 | TC05002126.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05029601.hg.1 | ENST00000335815.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05029601.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029600.hg.1 | TC05002126.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05029600.hg.1 | ENST00000335815.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05029600.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029604.hg.1 | TC05002126.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05029604.hg.1 | ENST00000335815.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05029604.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR05029608.hg.1 | TC05002126.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR05029608.hg.1 | ENST00000335815.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR05029608.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191158.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191158.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191158.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191158.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191158.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191158.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191158.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191158.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191158.hg.1 | TC0500013025.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500191160.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191160.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191160.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191160.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191160.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191160.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191160.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191160.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191160.hg.1 | TC0500013025.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500191156.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191156.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191156.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191156.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191156.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191156.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191156.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191156.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191156.hg.1 | TC0500013025.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500191154.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191154.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191154.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191154.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191154.hg.1 | TC0500013025.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500191159.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191159.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191159.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191159.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191159.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191159.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191159.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191159.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191159.hg.1 | TC0500013025.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500191162.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191162.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191162.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191162.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191162.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191162.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191162.hg.1 | TC0500013025.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000518048.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013025.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191155.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191155.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191155.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191155.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191155.hg.1 | ENST00000520331.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191155.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191155.hg.1 | TC0500013025.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0500191163.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191163.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191163.hg.1 | ENST00000335815.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0500191163.hg.1 | NM_005649.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0500191163.hg.1 | TC0500013025.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 214609 | - | - |
