Assay Details
| Gene Symbol: CLDN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006984.4 | NP_008915.1 | 1 - 2 | 277 | 82 | |
| GenBank mRNA | AK315737.1 | - | 1 - 2 | 229 | 82 | |
| BC010920.1 | - | 1 - 2 | 249 | 4246806 | 82 | |
| BU930613.1 | - | 267 | 82 | |||
| CR456845.1 | - | 1 - 2 | 216 | 82 | ||
| DA133712.1 | - | 1 - 2 | 277 | 82 | ||
| U89916.1 | - | 1 - 2 | 240 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000335.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13003471.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003471.hg.1 | TC13000335.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003472.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003472.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003472.hg.1 | TC13000335.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003474.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003474.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003474.hg.1 | ENST00000376873.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003474.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003474.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003474.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003474.hg.1 | TC13000335.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1300152234.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152234.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152234.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152234.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152234.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152234.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152234.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152234.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152234.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152234.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152234.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152234.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152234.hg.1 | TC1300007752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152230.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152230.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152230.hg.1 | TC1300007752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300152231.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152231.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152231.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152231.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152231.hg.1 | TC1300007752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300152232.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152232.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152232.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152232.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152232.hg.1 | TC1300007752.hg.1 | Clariom D TC |

CLDN10
Genomic Map